Assay Details
| Gene Symbol: CDK2AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122454.3 | XP_003122502.1 | 990 | 72 | ||
| GenBank mRNA | AK234345.1 | - | 1 - 2 | 667 | 72 | |
| AK396963.1 | - | 1 - 2 | 703 | 72 | ||
More Information
Set Membership: |
|

CDK2AP2
Genomic Map