Assay Details
| Gene Symbol: CDK2AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122454.3 | XP_003122502.1 | 1090 | 71 | ||
| XM_005660628.2 | XP_005660685.1 | 1310 | 71 | |||
| GenBank mRNA | AK234345.1 | - | 2 - 3 | 767 | 71 | |
| AK236753.1 | - | 2 - 3 | 401 | 71 | ||
| AK396780.1 | - | 1 - 2 | 1075 | 71 | ||
| AK396963.1 | - | 2 - 3 | 803 | 71 | ||
| AK400191.1 | - | 1 - 2 | 1056 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005660628.2 transcripts.
Set Membership: |
|

CDK2AP2
Genomic Map