Assay Details
| Gene Symbol: ZAR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065181.1 | XP_020920840.1 | 2409 | 118 | ||
| XM_021065182.1 | XP_020920841.1 | 849 | 118 | |||
| GenBank mRNA | AK234346.1 | - | 1 - 2 | 768 | 118 | |
More Information
Set Membership: |
|

ZAR1L
Genomic Map