Assay Details
| Gene Symbol: SMPD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929588.6 | XP_001929623.1 | 1396 | 59 | ||
| XM_001929589.6 | XP_001929624.1 | 2431 | 59 | |||
| XM_013981430.2 | XP_013836884.1 | 1427 | 59 | |||
| XM_013981435.2 | XP_013836889.1 | 1340 | 59 | |||
| GenBank mRNA | AK234371.1 | - | 13 - 14 | 1311 | 59 | |
| AK397600.1 | - | 12 - 13 | 2311 | 59 | ||
More Information
Set Membership: |
|

SMPD4
Genomic Map