Assay Details
| Gene Symbol: NPRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132226.4 | XP_003132274.1 | 1205 | 65 | ||
| XM_005669600.1 | XP_005669657.1 | 1280 | 65 | |||
| GenBank mRNA | AK234401.1 | - | 8 - 9 | 1648 | 65 | |
| AK234474.1 | - | 6 - 7 | 2187 | 65 | ||
| AY610117.1 | - | 10 - 11 | 1156 | 65 | ||
More Information
Set Membership: |
|

NPRL2
Genomic Map