Assay Details
| Gene Symbol: ADAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659689.3 | XP_005659746.1 | 832 | 77 | ||
| XM_013993096.2 | XP_013848550.1 | 877 | 77 | |||
| XM_013993097.2 | XP_013848551.1 | 646 | 77 | |||
| XM_013993098.2 | XP_013848552.1 | 832 | 77 | |||
| XM_013993099.1 | XP_013848553.1 | 592 | 77 | |||
| GenBank mRNA | AK234427.1 | - | 6 - 7 | 755 | 77 | |
More Information
Set Membership: |
|

ADAL
Genomic Map