Assay Details
| Gene Symbol: BACH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070882.1 | XP_020926541.1 | 4491 | 99 | ||
| XM_021070883.1 | XP_020926542.1 | 4905 | 99 | |||
| XM_021070884.1 | XP_020926543.1 | 4605 | 99 | |||
| XM_021070885.1 | XP_020926544.1 | 4312 | 99 | |||
| XM_021070886.1 | XP_020926545.1 | 3797 | 99 | |||
| XM_021070887.1 | XP_020926546.1 | 3669 | 99 | |||
| XM_021070888.1 | XP_020926547.1 | 3992 | 99 | |||
| XM_021070889.1 | XP_020926548.1 | 3622 | 99 | |||
| XM_021070890.1 | XP_020926549.1 | 3920 | 99 | |||
| XM_021070891.1 | XP_020926550.1 | 3926 | 99 | |||
| XM_021070892.1 | XP_020926551.1 | 3988 | 99 | |||
| GenBank mRNA | AK234585.1 | - | 1 | 816 | 99 | |
| AK400362.1 | - | 1 | 925 | 99 | ||
More Information
Set Membership: |
|

BACH1
Genomic Map