Assay Details
| Gene Symbol: PAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133921.6 | XP_003133969.3 | 1098 | 71 | ||
| XM_003483818.4 | XP_003483866.1 | 1261 | 71 | |||
| XM_005657615.3 | XP_005657672.1 | 809 | 71 | |||
| XM_005657616.3 | XP_005657673.1 | 939 | 71 | |||
| XM_005657617.3 | XP_005657674.1 | 825 | 71 | |||
| GenBank mRNA | AK234633.1 | - | 7 - 8 | 884 | 71 | |
More Information
Set Membership: |
|

PAIP1
Genomic Map