Assay Details
| Gene Symbol: CHST11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092628.1 | XP_020948287.1 | 104 | |||
| GenBank mRNA | AK234716.1 | - | 1 - 2 | 538 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CHST11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092628.1 | XP_020948287.1 | 104 | |||
| GenBank mRNA | AK234716.1 | - | 1 - 2 | 538 | 104 | |
Set Membership: |
|