Assay Details
| Gene Symbol: TMEM144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129006.6 | XP_003129054.3 | 1331 | 66 | ||
| XM_005666661.3 | XP_005666718.1 | 1375 | 66 | |||
| XM_013978685.2 | XP_013834139.1 | 1089 | 66 | |||
| XM_013978686.2 | XP_013834140.1 | 1091 | 66 | |||
| XM_013978687.2 | XP_013834141.1 | 1252 | 66 | |||
| XM_021101288.1 | XP_020956947.1 | 1140 | 66 | |||
| GenBank mRNA | AK234800.1 | - | 10 - 11 | 1331 | 66 | |
| AK236252.1 | - | 10 - 11 | 1219 | 66 | ||
| AK393067.1 | - | 11 - 12 | 1269 | 66 | ||
| AK398828.1 | - | 11 - 12 | 1252 | 66 | ||
More Information
Set Membership: |
|

TMEM144
Genomic Map