Assay Details
| Gene Symbol: MPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135524.4 | XP_003135572.1 | 971 | 58 | ||
| XM_021080526.1 | XP_020936185.1 | 817 | 58 | |||
| GenBank mRNA | AK234821.1 | - | 7 - 8 | 785 | 58 | |
| AK392380.1 | - | 7 - 8 | 909 | 58 | ||
| AK399182.1 | - | 7 - 8 | 938 | 58 | ||
More Information
Set Membership: |
|

MPP1
Genomic Map