Assay Details
| Gene Symbol: GALK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131193.2 | XP_003131241.1 | 1033 | 81 | ||
| GenBank mRNA | AK234860.1 | - | 6 - 7 | 1000 | 81 | |
| AK393980.1 | - | 6 - 7 | 1315 | 81 | ||
More Information
Set Membership: |
|

GALK1
Genomic Map