Assay Details
| Gene Symbol: AGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927332.7 | XP_001927367.2 | 1319 | 71 | ||
| GenBank mRNA | AK234888.1 | - | 1 - 2 | 192 | 71 | |
| AK350203.1 | - | 1 - 2 | 121 | 71 | ||
| AY610020.1 | - | 1 - 2 | 219 | 71 | ||
More Information
Set Membership: |
|

AGA
Genomic Map