Assay Details
| Gene Symbol: YARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356295.4 | XP_003356343.3 | 263 | 105 | ||
| XM_013999119.2 | XP_013854573.1 | 641 | 105 | |||
| XM_021095817.1 | XP_020951476.1 | 695 | 105 | |||
| GenBank mRNA | AK234892.1 | - | 2 - 3 | 316 | 105 | |
| AK239477.1 | - | 2 - 3 | 316 | 105 | ||
| AK393114.1 | - | 2 - 3 | 316 | 105 | ||
| AK399965.1 | - | 2 - 3 | 292 | 105 | ||
| AY610116.1 | - | 2 - 3 | 264 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013999119.2, XM_021095817.1 transcripts.
Set Membership: |
|

YARS1
Genomic Map