Assay Details
Gene Symbol: ERGIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003134052.6 | XP_003134100.4 | 669 | 63 | ||
XM_013990648.2 | XP_013846102.1 | 720 | 63 | |||
GenBank mRNA | AK235006.1 | - | 1 - 2 | 35 | 63 | |
AK395943.1 | - | 7 - 8 | 589 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
