Assay Details
Gene Symbol: SMIM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003362205.4 | XP_003362253.1 | 202 | 62 | ||
GenBank mRNA | AK235103.1 | - | 1 - 2 | 73 | 62 | |
AK349038.1 | - | 1 - 2 | 208 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003362205.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
