Assay Details
| Gene Symbol: TSEN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013979927.2 | XP_013835381.2 | 200 | 76 | ||
| XM_013979928.2 | XP_013835382.2 | 225 | 76 | |||
| GenBank mRNA | AK235130.1 | - | 1 - 2 | 141 | 76 | |
More Information
Set Membership: |
|

TSEN15
Genomic Map