Assay Details
| Gene Symbol: CASC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067024.1 | XP_020922683.1 | 2160 | 62 | ||
| GenBank mRNA | AK235140.1 | - | 1 - 2 | 224 | 62 | |
| AK350283.1 | - | 2 - 3 | 139 | 62 | ||
More Information
Set Membership: |
|

CASC3
Genomic Map