Assay Details
| Gene Symbol: DDAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021098458.1 | XP_020954117.1 | 688 | 58 | ||
| XM_021098459.1 | XP_020954118.1 | 1100 | 58 | |||
| XM_021098460.1 | XP_020954119.1 | 1047 | 58 | |||
| GenBank mRNA | AK235171.1 | - | 3 - 4 | 673 | 58 | |
| KP696756.1 | - | 2 - 3 | 398 | 58 | ||
More Information
Set Membership: |
|

DDAH2
Genomic Map