Assay Details
| Gene Symbol: VPS26C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483358.4 | XP_003483406.1 | 4561 | 94 | ||
| XM_005670333.3 | XP_005670390.1 | 4584 | 94 | |||
| GenBank mRNA | AK235177.1 | - | 1 | 1417 | 94 | |
More Information
Set Membership: |
|

VPS26C
Genomic Map