Assay Details
| Gene Symbol: KCTD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005670738.3 | XP_005670795.2 | 233 | 64 | ||
| XM_013982975.2 | XP_013838429.1 | 233 | 64 | |||
| GenBank mRNA | AK235200.1 | - | 1 - 2 | 44 | 64 | |
| AK343264.1 | - | 1 - 2 | 39 | 64 | ||
More Information
Set Membership: |
|

KCTD10
Genomic Map