Assay Details
| Gene Symbol: AMDHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013987848.2 | XP_013843302.1 | 1134 | 58 | ||
| XM_013987849.2 | XP_013843303.1 | 1143 | 58 | |||
| XM_021086752.1 | XP_020942411.1 | 1206 | 58 | |||
| GenBank mRNA | AK235211.1 | - | 9 - 10 | 1087 | 58 | |
| AK238502.1 | - | 9 - 10 | 1114 | 58 | ||
| AK398425.1 | - | 9 - 10 | 1121 | 58 | ||
More Information
Set Membership: |
|

AMDHD2
Genomic Map