Assay Details
| Gene Symbol: DENND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021062259.1 | XP_020917918.1 | 2579 | 101 | ||
| GenBank mRNA | AK235216.1 | - | 4 - 5 | 628 | 101 | |
More Information
Set Membership: |
|

DENND2B
Genomic Map