Assay Details
| Gene Symbol: PLEKHA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005673281.2 | XP_005673338.1 | 1212 | 88 | ||
| XM_013985757.2 | XP_013841211.1 | 1161 | 88 | |||
| XM_021079155.1 | XP_020934814.1 | 775 | 88 | |||
| XM_021079156.1 | XP_020934815.1 | 1161 | 88 | |||
| GenBank mRNA | AK235226.1 | - | 1 - 2 | 145 | 88 | |
| AK392454.1 | - | 7 - 8 | 1167 | 88 | ||
More Information
Set Membership: |
|

PLEKHA8
Genomic Map