Assay Details
| Gene Symbol: SEPTIN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129096.6 | XP_003129144.1 | 332 | 60 | ||
| XM_005666781.3 | XP_005666838.1 | 299 | 60 | |||
| XM_005666782.3 | XP_005666839.1 | 299 | 60 | |||
| XM_005666785.3 | XP_005666842.1 | 299 | 60 | |||
| XM_005666786.3 | XP_005666843.1 | 299 | 60 | |||
| XM_013978808.2 | XP_013834262.1 | 299 | 60 | |||
| XM_021101469.1 | XP_020957128.1 | 260 | 60 | |||
| XM_021101471.1 | XP_020957130.1 | 259 | 60 | |||
| XM_021101472.1 | XP_020957131.1 | 260 | 60 | |||
| XM_021101473.1 | XP_020957132.1 | 260 | 60 | |||
| XM_021101474.1 | XP_020957133.1 | 363 | 60 | |||
| XM_021101476.1 | XP_020957135.1 | 363 | 60 | |||
| GenBank mRNA | AK235359.1 | - | 2 - 3 | 286 | 60 | |
| AK390400.1 | - | 2 - 3 | 291 | 60 | ||
| KU358544.1 | - | 3 - 4 | 189 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021101474.1, XM_021101476.1 transcripts.
Set Membership: |
|

SEPTIN11
Genomic Map