Assay Details
| Gene Symbol: EMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358517.4 | XP_003358565.1 | 353 | 66 | ||
| GenBank mRNA | AK235371.1 | - | 1 - 2 | 341 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: EMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358517.4 | XP_003358565.1 | 353 | 66 | ||
| GenBank mRNA | AK235371.1 | - | 1 - 2 | 341 | 66 | |
Set Membership: |
|