Assay Details
| Gene Symbol: NAP1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003360391.4 | XP_003360439.1 | 2232 | 96 | ||
| GenBank mRNA | AK235381.1 | - | 1 | 2216 | 100 | |
| AK394907.1 | - | 1 | 2223 | 96 | ||
| AK395265.1 | - | 1 | 2156 | 99 | ||
More Information
Set Membership: |
|

NAP1L3
Genomic Map