Assay Details
| Gene Symbol: CRYBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095628.1 | XP_020951287.1 | 5201 | 98 | ||
| GenBank mRNA | AK235383.1 | - | 5 - 6 | 696 | 98 | |
More Information
Set Membership: |
|

CRYBG2
Genomic Map