Assay Details
| Gene Symbol: PCBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125057.4 | XP_003125105.1 | 966 | 66 | ||
| GenBank mRNA | AK235462.1 | - | 1 | 822 | 66 | |
| AK397353.1 | - | 1 | 895 | 66 | ||
| AK397422.1 | - | 1 | 823 | 66 | ||
| AK398850.1 | - | 1 | 680 | 66 | ||
More Information
Set Membership: |
|

PCBP1
Genomic Map