Assay Details
| Gene Symbol: RAB23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929636.4 | XP_001929671.1 | 3593 | 129 | ||
| XM_021098560.1 | XP_020954219.1 | 3574 | 129 | |||
| GenBank mRNA | AK235662.1 | - | 1 | 822 | 129 | |
More Information
Set Membership: |
|

RAB23
Genomic Map