Assay Details
| Gene Symbol: ACP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928346.6 | XP_001928381.3 | 1034 | 56 | ||
| XM_021089952.1 | XP_020945611.1 | 947 | 56 | |||
| GenBank mRNA | AK235757.1 | - | 4 - 5 | 996 | 56 | |
| AK351385.1 | - | 4 - 5 | 996 | 56 | ||
| AY609695.1 | - | 4 - 5 | 866 | 56 | ||
More Information
Set Membership: |
|

ACP6
Genomic Map