Assay Details
| Gene Symbol: LOC110255277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021074730.1 | XP_020930389.1 | 1372 | 94 | ||
| GenBank mRNA | AB613817.1 | - | 9 - 10 | 1203 | 94 | |
More Information
Set Membership: |
|

LOC110255277
Genomic Map