Assay Details
| Gene Symbol: ALPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097681.1 | XP_020953340.1 | 1261 | 69 | ||
| XM_021097682.1 | XP_020953341.1 | 1276 | 69 | |||
| XM_021097683.1 | XP_020953342.1 | 1279 | 69 | |||
| XM_021097684.1 | XP_020953343.1 | 1199 | 69 | |||
| XM_021097685.1 | XP_020953344.1 | 1275 | 69 | |||
| XM_021097686.1 | XP_020953345.1 | 1263 | 69 | |||
| GenBank mRNA | AH012163.2 | - | 8 - 9 | 986 | 69 | |
| AK398634.1 | - | 4 - 5 | 436 | 69 | ||
More Information
Set Membership: |
|

ALPL
Genomic Map