Assay Details
| Gene Symbol: SCPEP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656968.3 | XP_005657025.1 | 1199 | 66 | ||
| GenBank mRNA | AK230482.1 | - | 11 - 12 | 1137 | 66 | |
| AK237259.1 | - | 11 - 12 | 1123 | 66 | ||
| AK391396.1 | - | 11 - 12 | 1178 | 66 | ||
| AK397438.1 | - | 11 - 12 | 1124 | 66 | ||
| AK399585.1 | - | 11 - 12 | 1178 | 66 | ||
More Information
Set Membership: |
|

SCPEP1
Genomic Map