Assay Details
| Gene Symbol: ADGRE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083689.1 | XP_020939348.1 | 187 | 99 | ||
| GenBank mRNA | AK230572.1 | - | 2 - 3 | 152 | 99 | |
| AK230702.1 | - | 2 - 3 | 124 | 99 | ||
| AK230812.1 | - | 2 - 3 | 108 | 99 | ||
More Information
Set Membership: |
|

ADGRE3
Genomic Map