Assay Details
| Gene Symbol: ETNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092211.1 | XP_020947870.1 | 6473 | 95 | ||
| GenBank mRNA | AK230574.1 | - | 1 | 1923 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ETNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092211.1 | XP_020947870.1 | 6473 | 95 | ||
| GenBank mRNA | AK230574.1 | - | 1 | 1923 | 95 | |
Set Membership: |
|