Assay Details
| Gene Symbol: STX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926476.4 | XP_001926511.1 | 820 | 72 | ||
| GenBank mRNA | AK230656.1 | - | 8 - 9 | 768 | 72 | |
| AK395138.1 | - | 8 - 9 | 776 | 72 | ||
| AK399470.1 | - | 8 - 9 | 768 | 72 | ||
| AY610058.1 | - | 8 - 9 | 734 | 72 | ||
More Information
Set Membership: |
|

STX7
Genomic Map