Assay Details
| Gene Symbol: EXOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132105.6 | XP_003132153.5 | 1386 | 70 | ||
| XM_021071674.1 | XP_020927333.1 | 1387 | 70 | |||
| XM_021071675.1 | XP_020927334.1 | 764 | 70 | |||
| GenBank mRNA | AK230662.1 | - | 4 - 5 | 590 | 70 | |
| AK237071.1 | - | 5 - 6 | 677 | 70 | ||
| AK238567.1 | - | 5 - 6 | 662 | 70 | ||
| AK343681.1 | - | 4 - 5 | 594 | 70 | ||
| AK345328.1 | - | 1 - 2 | 186 | 70 | ||
| AK400166.1 | - | 5 - 6 | 674 | 70 | ||
More Information
Set Membership: |
|

EXOG
Genomic Map