Assay Details
| Gene Symbol: IFI44L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127919.4 | XP_003127967.3 | 1307 | 140 | ||
| GenBank mRNA | AK230677.1 | - | 7 - 8 | 1258 | 140 | |
| AK230699.1 | - | 7 - 8 | 1258 | 140 | ||
| AK389881.1 | - | 7 - 8 | 1258 | 140 | ||
More Information
Set Membership: |
|

IFI44L
Genomic Map