Assay Details
| Gene Symbol: YY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099699.1 | XP_020955358.1 | 770 | 82 | ||
| GenBank mRNA | AK230712.1 | - | 1 - 2 | 91 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: YY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099699.1 | XP_020955358.1 | 770 | 82 | ||
| GenBank mRNA | AK230712.1 | - | 1 - 2 | 91 | 82 | |
Set Membership: |
|