Assay Details
| Gene Symbol: SLC35A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353254.5 | XP_003353302.1 | 632 | 76 | ||
| XM_021089797.1 | XP_020945456.1 | 480 | 76 | |||
| GenBank mRNA | AK230731.1 | - | 3 - 4 | 379 | 76 | |
More Information
Set Membership: |
|

SLC35A1
Genomic Map