Assay Details
| Gene Symbol: TENT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927710.5 | XP_001927745.1 | 4939 | 81 | ||
| XM_013992805.2 | XP_013848259.1 | 5020 | 81 | |||
| GenBank mRNA | AK230737.1 | - | 1 | 2013 | 81 | |
More Information
Set Membership: |
|

TENT5A
Genomic Map