Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003483513.4 | XP_003483561.2 | 3506 | 78 | ||
XM_021073589.1 | XP_020929248.1 | 2046 | 78 | |||
XM_021073591.1 | XP_020929250.1 | 2090 | 78 | |||
XM_021073592.1 | XP_020929251.1 | 2153 | 78 | |||
XM_021073593.1 | XP_020929252.1 | 2047 | 78 | |||
XM_021073594.1 | XP_020929253.1 | 2077 | 78 | |||
GenBank mRNA | AK230746.1 | - | 6 - 7 | 668 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
