Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003483513.4 | XP_003483561.2 | 3707 | 93 | ||
XM_021073589.1 | XP_020929248.1 | 2247 | 93 | |||
XM_021073591.1 | XP_020929250.1 | 2291 | 93 | |||
XM_021073592.1 | XP_020929251.1 | 2354 | 93 | |||
XM_021073593.1 | XP_020929252.1 | 2248 | 93 | |||
XM_021073594.1 | XP_020929253.1 | 2278 | 93 | |||
XM_021073595.1 | XP_020929254.1 | 1918 | 93 | |||
GenBank mRNA | AK230746.1 | - | 7 - 8 | 869 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
