Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003483513.4 | XP_003483561.2 | 3940 | 96 | ||
XM_021073589.1 | XP_020929248.1 | 2480 | 96 | |||
XM_021073591.1 | XP_020929250.1 | 2524 | 96 | |||
XM_021073592.1 | XP_020929251.1 | 2587 | 96 | |||
XM_021073593.1 | XP_020929252.1 | 2481 | 96 | |||
XM_021073594.1 | XP_020929253.1 | 2511 | 96 | |||
XM_021073595.1 | XP_020929254.1 | 2151 | 96 | |||
GenBank mRNA | AK230746.1 | - | 8 - 9 | 1102 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
