Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003483513.4 | XP_003483561.2 | 4052 | 69 | ||
XM_021073589.1 | XP_020929248.1 | 2592 | 69 | |||
XM_021073591.1 | XP_020929250.1 | 2636 | 69 | |||
XM_021073592.1 | XP_020929251.1 | 2699 | 69 | |||
XM_021073595.1 | XP_020929254.1 | 2263 | 69 | |||
GenBank mRNA | AK230746.1 | - | 9 - 10 | 1214 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
