Assay Details
| Gene Symbol: ANXA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129218.5 | XP_003129266.2 | 961 | 69 | ||
| GenBank mRNA | AK230760.1 | - | 8 - 9 | 625 | 69 | |
| AK234913.1 | - | 8 - 9 | 692 | 69 | ||
| AK239527.1 | - | 8 - 9 | 662 | 69 | ||
| AK389690.1 | - | 8 - 9 | 671 | 69 | ||
| AK397574.1 | - | 8 - 9 | 691 | 69 | ||
| AK399468.1 | - | 8 - 9 | 679 | 69 | ||
| AK399561.1 | - | 8 - 9 | 679 | 69 | ||
| AK399872.1 | - | 8 - 9 | 724 | 69 | ||
More Information
Set Membership: |
|

ANXA5
Genomic Map