Assay Details
| Gene Symbol: RPP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924771.6 | XP_001924806.1 | 952 | 81 | ||
| GenBank mRNA | AK230820.1 | - | 10 - 11 | 709 | 81 | |
More Information
Set Membership: |
|

RPP30
Genomic Map