Assay Details
| Gene Symbol: DDX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066758.1 | XP_020922417.1 | 1770 | 137 | ||
| XM_021066760.1 | XP_020922419.1 | 1703 | 137 | |||
| XM_021066761.1 | XP_020922420.1 | 1425 | 137 | |||
| XM_021066762.1 | XP_020922421.1 | 1307 | 137 | |||
| GenBank mRNA | AK230829.1 | - | 10 - 11 | 1289 | 137 | |
| AK235519.1 | - | 11 - 12 | 1390 | 137 | ||
| AK239448.1 | - | 12 - 13 | 1468 | 137 | ||
| AK350880.1 | - | 12 - 13 | 1475 | 137 | ||
| AK396121.1 | - | 11 - 12 | 1386 | 137 | ||
| AK399667.1 | - | 12 - 13 | 1515 | 137 | ||
| KM434334.1 | - | 11 - 12 | 1216 | 137 | ||
More Information
Set Membership: |
|

DDX5
Genomic Map