Assay Details
| Gene Symbol: TAF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095677.1 | XP_020951336.1 | 642 | 69 | ||
| XM_021095678.1 | XP_020951337.1 | 574 | 69 | |||
| XM_021095679.1 | XP_020951338.1 | 990 | 69 | |||
| GenBank mRNA | AK230834.1 | - | 4 - 5 | 483 | 69 | |
| AK343725.1 | - | 4 - 5 | 483 | 69 | ||
| AK397226.1 | - | 4 - 5 | 541 | 69 | ||
More Information
Set Membership: |
|

TAF12
Genomic Map